(R)-tert-Butyl (4-oxobutan-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A101142-100MG A101142-250MG A101142-25MG A101142-1G
103795-820EA 277 USD
103795-820 103821-510 77188-546 77188-548 77188-550 77296-996
(R)-tert-Butyl (4-oxobutan-2-yl)carbamate 97%
(R)-tert-Butyl (4-oxobutan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
Boiling Pt: 279.9±23.0 °C (760 mm Hg)
Flash Pt: 123.1 °C
Storage Temperature: Freezer
MDL Number: MFCD18835622
CAS Number: 497861-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR