(R)-tert-Butyl (4-hydroxybutan-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A292400-1G A292400-5G A292400-10G A292400-250MG A292400-25G
103813-938EA 50 USD
103813-938 103835-078 76837-678 76837-680 77099-346 77099-348 77099-350
(R)-tert-Butyl (4-hydroxybutan-2-yl)carbamate 95%
(R)-tert-Butyl (4-hydroxybutan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MW: 189.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18831346
CAS Number: 167216-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR