(R)-tert-Butyl (4-cyano-2,3-dihydro-1H-inden-1-yl)carbamate 97%

Supplier: Ambeed
Warning

A185292-250MG A185292-1G
103815-840EA 155 USD
103815-840 77260-358 77301-964
(R)-tert-Butyl (4-cyano-2,3-dihydro-1H-inden-1-yl)carbamate 97%
(R)-tert-Butyl (4-cyano-2,3-dihydro-1H-inden-1-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₂
MW: 258.31 g/mol
Boiling Pt: 357.7±37.0 °C (760 mm Hg)
Flash Pt: 170.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD28502614
CAS Number: 1306763-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR