(R)-tert-Butyl(2-oxooxetan-3-yl)carbamate 95%
Supplier: Ambeed
|
Warning
|
A549770-100MG
A549770-10G
A549770-1G
A549770-250MG
103799-680EA
22
USD
103799-680
103839-814
76696-114
76696-116
76696-118
76696-120
(R)-tert-Butyl(2-oxooxetan-3-yl)carbamate 95%
(R)-tert-Butyl(2-oxooxetan-3-yl)carbamate
Precaución: for Research Use Only
Formula:
C₈H₁₃NO₄ Boiling Pt: 319 °C (760 mm Hg) Melting Pt: 121…122 °C Flash Pt: 147 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11519129 CAS Number: 126330-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...