(R)-tert-Butyl (1-(4-bromophenyl)ethyl)carbamate 98%

Supplier: Ambeed
Warning

A143099-100MG A143099-250MG
103803-238EA 105 USD
103803-238 77027-676 77027-678
(R)-tert-Butyl (1-(4-bromophenyl)ethyl)carbamate 98%
(R)-tert-Butyl (1-(4-bromophenyl)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₂
MW: 300.19 g/mol
Boiling Pt: 383.1 °C (760 mm Hg)
Melting Pt: 131…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11506010
CAS Number: 578729-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR