D-β-Proline 95%

Supplier: Ambeed
Warning

A133874-10G A133874-250MG A133874-1G A133874-5G A133874-25G
103801-924EA 24 USD
103801-924 76782-504 76782-506 77314-620 77314-622 77666-930
D-β-Proline 95%
D-beta-Proline

Precaución: for Research Use Only

Formula: C₅H₉NO₂
Flash Pt: 106.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08272856
CAS Number: 72580-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR