(R)-Pyrrolidin-3-ylmethanol 97%

Supplier: Ambeed
Warning

A139279-10G A139279-250MG A139279-1G
103802-726EA 24 USD
103802-726 77137-240 77137-242 77299-072
(R)-Pyrrolidin-3-ylmethanol 97%
(R)-Pyrrolidin-3-ylmethanol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 176.1 °C (760 mm Hg)
Flash Pt: 89.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09607969
CAS Number: 110013-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR