(R)-Pentan-2-amine hydrochloride 97%

Supplier: Ambeed
Warning

A130183-100MG A130183-250MG A130183-50MG A130183-1G
103801-388EA 208 USD
103801-388 77246-500 77246-502 77246-504 77298-394
(R)-Pentan-2-amine hydrochloride 97%
(R)-Pentan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
MW: 123.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28501705
CAS Number: 101689-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR