(R)-(+)-N,α-Dimethylbenzylamine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-N-Methyl-1-phenylethanamine, (R)-N-Methyl-alpha-phenylethylamine

A101192-25G A101192-1G A101192-5G
103795-850EA 57 USD
103795-850 103821-534 77245-102 77282-674 77297-016
(R)-(+)-N,α-Dimethylbenzylamine 97%
(R)-(+)-N,α-Dimethylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.2062 g/mol
Boiling Pt: 178.7 °C (760 mm Hg)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00044966
CAS Number: 5933-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR