(R)-(+)-N-Benzyl-α-methylbenzylamine 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
(+)-Benzylphenethylamine, (R)-N-Benzyl-1-phenylethanamine, (R)-(+)-N-Benzyl-1-phenylethylamine
A252635-100G
A252635-10G
A252635-25G
A252635-5G
A252635-500G
103809-798EA
298
USD
103809-798
103851-898
76976-712
76976-714
77091-318
77304-586
77601-034
(R)-(+)-N-Benzyl-α-methylbenzylamine 96%
(R)-(+)-N-Benzyl-α-methylbenzylamine
Precaución: for Research Use Only
Formula:
C₁₅H₁₇N MW: 211.31 g/mol Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015010 CAS Number: 38235-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...