(R)-(-)-1-Boc-3-hydroxypyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-N-Boc-3-pyrrolidinol, tert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate, (R)-N-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine

A121392-100G A121392-10G A121392-1G A121392-25G A121392-500G A121392-5G A121392-1000G
77258-108EA 76.22 USD
77258-108 77258-110 77258-112 77258-114 77258-116 77312-186 77745-893
(R)-(-)-1-Boc-3-hydroxypyrrolidine 97%
(R)-(-)-1-Boc-3-hydroxypyrrolidine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
Melting Pt: 62…65 °C
Flash Pt: 119.1 °C
Storage Temperature: Ambient
MDL Number: MFCD01317838
CAS Number: 109431-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR