(R)-Morpholin-3-ylmethanol hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3R)-3-(Hydroxymethyl)morpholine hydrochloride
A201508-100MG
A201508-250MG
A201508-1G
A201508-5G
103817-302EA
14
USD
103817-302
103847-942
76814-310
76814-312
77302-758
77302-760
(R)-Morpholin-3-ylmethanol hydrochloride 95%
(R)-Morpholin-3-ylmethanol hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO₂ MW: 153.6100 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09910295 CAS Number: 1212377-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...