(R)-Methyl pyrrolidine-3-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A109363-100MG A109363-250MG A109363-1G A109363-5G
103797-500EA 33 USD
103797-500 103823-296 76781-008 76781-010 77310-112 77310-114
(R)-Methyl pyrrolidine-3-carboxylate hydrochloride 95%
(R)-Methyl pyrrolidine-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂·ClH
MW: 165.618 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11840195
CAS Number: 874964-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR