(R)-(-)-Methyl-3-hydroxybutyrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-Methyl 3-hydroxybutanoate, Methyl (R)-3-hydroxybutyrate, Methyl (R)-(-)-3-Hydroxybutyrate
A233720-100G
A233720-10G
A233720-500G
A233720-25G
A233720-5G
A233720-1000G
103850-664EA
5
USD
103850-664
76801-132
76801-134
76801-136
77089-636
77089-638
77667-930
(R)-(-)-Methyl-3-hydroxybutyrate 97%
(R)-(-)-Methyl-3-hydroxybutyrate
Precaución: for Research Use Only
Formula:
C₅H₁₀O₃ Boiling Pt: 161.9 °C (760 mm Hg) Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063289 CAS Number: 3976-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...