(R)-Methyl 3-((tert-butoxycarbonyl)amino)butanoate 97%

Supplier: Ambeed
Warning

A266051-10G A266051-250MG A266051-1G A266051-5G
103812-778EA 86 USD
103812-778 103852-676 76722-946 76722-948 77092-374 77092-376
(R)-Methyl 3-((tert-butoxycarbonyl)amino)butanoate 97%
(R)-Methyl 3-((tert-butoxycarbonyl)amino)butanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03788463
CAS Number: 159877-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR