Methyl D-3-phenyllactate 96%

Supplier: Ambeed
Warning

Synonyms: D-3-Phenyllactic Acid Methyl Ester, (R)-Methyl 2-hydroxy-3-phenylpropanoate, Methyl (R)-2-Hydroxy-3-phenylpropionate, (R)-2-Hydroxy-3-phenylpropionic Acid Methyl Ester

A290402-1G A290402-25G A290402-5G A290402-100G A290402-10G A290402-250MG
103813-864EA 7 USD
103813-864 103835-006 76837-582 76837-584 76837-586 76977-566 76977-568 76977-570
Methyl D-3-phenyllactate 96%
Methyl D-3-phenyllactate

Precaución: For research use only

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Boiling Pt: 286 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09842268
CAS Number: 27000-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR