D-Phenylalanine methyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Methyl 2-amino-3-phenylpropanoate hydrochloride, H-D-Phe-OMe.HCl, H-D-Phe-OMe·HCl

A138087-100G A138087-10G A138087-500G A138087-25G A138087-5G
76688-896EA 45 USD
76688-896 76688-898 76688-900 77315-558 77315-560
D-Phenylalanine methyl ester hydrochloride 98%
D-Phenylalanine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
Melting Pt: 159…163 °C
Flash Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066112
CAS Number: 13033-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR