(R)-Methyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A151115-250MG
A151115-1G
103804-234EA
102
USD
103804-234
103842-380
77247-264
77299-850
(R)-Methyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride 97%
(R)-Methyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₆ClNO₄S MW: 293.77 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30570303 CAS Number: 2049127-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...