(R)-Methyl 2,3-dihydroxypropanoate 97%

Supplier: Ambeed
Warning

A227282-1G A227282-100MG A227282-10G A227282-250MG
103819-310EA 19 USD
103819-310 103850-216 77088-982 77165-438 77165-440 77165-442
(R)-Methyl 2,3-dihydroxypropanoate 97%
(R)-Methyl 2,3-dihydroxypropanoate

Precaución: for Research Use Only

Formula: C₄H₈O₄
Boiling Pt: 140 °C (15 mm Hg)
Flash Pt: 103.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06204263
CAS Number: 18289-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR