Methyl (R)-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: (+)-2,3-O-Isopropylidene-D-glyceric Acid Methyl Ester, (R)-Methyl 2,2-dimethyl-1,3-dioxolane-4-carboxylate, (+)-Methyl 2,3-O-Isopropylidene-D-glycerate, (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester

A116621-100G A116621-10G A116621-25G A116621-1G
103798-770EA 5 USD
103798-770 103824-634 76971-228 76971-230 76971-232 77311-432
Methyl (R)-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate 95%
Methyl (R)-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00066527
CAS Number: 52373-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR