(R)-(-)-Ethyl-3-piperidinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl (R)-(-)-3-Piperidinecarboxylate, (R)-Ethyl piperidine-3-carboxylate

A162559-1G A162559-5G A162559-10G A162559-100G
103805-628EA 9 USD
103805-628 103843-878 77319-926 77319-928 77611-582 77881-267
(R)-(-)-Ethyl-3-piperidinecarboxylate 98%
(R)-(-)-Ethyl 3-piperidinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MW: 157.2102 g/mol
Boiling Pt: 238.6 °C (760 mm Hg)
Flash Pt: 104 °C
Storage Temperature: Freezer
MDL Number: MFCD03093637
CAS Number: 25137-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR