H-D-Glu(OtBu)-OtBu·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Glu(OtBu)-OtBu·HCl, (R)-Di-tert-butyl 2-aminopentanedioate hydrochloride

A133931-100G A133931-10G A133931-250MG A133931-5G A133931-1G A133931-25G
103801-934EA 6 USD
103801-934 76797-976 76797-978 76797-980 77298-664 77314-632 77314-634
H-D-Glu(OtBu)-OtBu·HCl 97%
H-D-Glu(OtBu)-OtBu·HCl

Precaución: for Research Use Only

Formula: C₁₃H₂₆ClNO₄
MW: 295.804 g/mol
Melting Pt: 125…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237068
CAS Number: 172793-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR