(R)-Dimethylpyrrolidin-3-ylamine dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Dimethylpyrrolidin-3-yl-amine dihydrochloride

A263535-1G A263535-5G A263535-250MG
103812-628EA 43 USD
103812-628 103852-516 77092-150 77092-152 77140-788
(R)-Dimethylpyrrolidin-3-ylamine dihydrochloride 98%
(R)-Dimethylpyrrolidin-3-ylamine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
Melting Pt: 194…197 °C
Storage Temperature: Ambient
MDL Number: MFCD08704347
CAS Number: 864448-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR