(R)-Boroleucine-(1S,2S,3R,5S)-(+)-pinanediol ester trifluoroacetate 97%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Methyl-1-((3aS,4S,6S,7aR)-3a,5,5-trimethylhexahydro-4,6-methanobenzo[d][1,3,2]dioxaborol-2-yl)butan-1-amine 2,2,2-trifluoroacetate, Bortezomib Amino R Isomer (TFA salt), (R)-BoroLeu-(+)-Pinanediol trifluoroacetate

A192536-1G A192536-25G A192536-10G A192536-250MG
103816-452EA 15 USD
103816-452 103847-060 77084-936 77084-938 77139-106 77139-108
(R)-Boroleucine-(1S,2S,3R,5S)-(+)-pinanediol ester trifluoroacetate 97%
(R)-Boroleucine-(1S,2S,3R,5S)-(+)-pinanediol ester trifluoroacetate

Precaución: for Research Use Only

Formula: C₁₇H₂₉BF₃NO₄
MW: 379.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566030
CAS Number: 179324-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR