(R)-1-Cbz-3-aminopiperidine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-Benzyl 3-aminopiperidine-1-carboxylate

A102322-100MG A102322-250MG A102322-5G
103796-358EA 19 USD
103796-358 103822-050 77025-202 77025-204 77308-936
(R)-1-Cbz-3-aminopiperidine 97%
(R)-1-Cbz-3-aminopiperidine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.294 g/mol
Boiling Pt: 367.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD05864625
CAS Number: 1044560-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR