(R)-Benzyl(5-oxotetrahydrofuran-3-yl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Benzyl (5-oxotetrahydrofuran-3-yl)carbamate, (R)-4-(Benzyloxycarbonylamino)tetrahydrofuran-2-one, (R)-beta-(Cbz-amino)-gamma-butyrolactone, (R)-beta-(Benzyloxycarbonylamino)-gamma-butyrolactone, (R)-beta-(Carbobenzoxyamino)-gamma-butyrolactone, (R)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one, (R)-4-(Cbz-amino)tetrahydrofuran-2-one, Benzyl (R)-Tetrahydro-5-oxo-3-furanylcarbamate

A124062-100G A124062-100MG A124062-10G A124062-250MG A124062-25G A124062-1G A124062-5G
103800-480EA 13 USD
103800-480 77136-636 77136-638 77136-640 77136-642 77136-644 77312-608 77312-610
(R)-Benzyl(5-oxotetrahydrofuran-3-yl)carbamate 98%
(R)-Benzyl(5-oxotetrahydrofuran-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₄
MW: 235.2400 g/mol
Boiling Pt: 466.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09839299
CAS Number: 118399-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR