(R)-Azetidine-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A169843-10G
A169843-250MG
A169843-5G
A169843-1G
A169843-25G
103806-420EA
20
USD
103806-420
103844-684
76824-456
76824-458
77081-780
77301-004
77301-006
(R)-Azetidine-2-carboxylic acid 95%
(R)-Azetidine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₄H₇NO₂ MW: 101.1039 g/mol Boiling Pt: 242 °C (760 mm Hg) Melting Pt: 209…211 °C Flash Pt: 100.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00800731 CAS Number: 7729-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...