(R)-6-(4-Aminophenyl)-5-methyl-4,5-dihydropyridazin-3(2H)-one 97%

Supplier: Ambeed
Warning

Synonyms: OR-1855

A120422-250MG A120422-1G A120422-100MG
103799-928EA 91 USD
103799-928 76766-426 77312-034 77624-000
(R)-6-(4-Aminophenyl)-5-methyl-4,5-dihydropyridazin-3(2H)-one 97%
(R)-6-(4-Aminophenyl)-5-methyl-4,5-dihydropyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 206…208 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09032921
CAS Number: 101328-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR