(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-5-Bromo-3-(1-(2,6-dichloro-3-fluorophenyl)ethoxy)pyridin-2-amine

A167135-10G A167135-250MG A167135-25G A167135-1G A167135-5G
103806-134EA 34 USD
103806-134 103844-396 76872-236 76872-238 76872-240 77320-686 77320-688
(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine 98%
(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrCl₂FN₂O
MW: 380.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18207061
CAS Number: 877399-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR