(R)-5-(Aminomethyl)pyrrolidin-2-one hydrochloride 96%
Supplier: Ambeed
|
Warning
|
A279626-100MG
A279626-250MG
A279626-5G
103813-474EA
63
USD
103813-474
76988-460
76988-462
76988-464
(R)-5-(Aminomethyl)pyrrolidin-2-one hydrochloride 96%
(R)-5-(Aminomethyl)pyrrolidin-2-one hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₁ClN₂O MW: 150.61 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD26743433 CAS Number: 1956434-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...