(R)-5-(2,2-Dimethyl-4H-benzo[d][1,3]dioxin-6-yl)oxazolidin-2-one 97%

Supplier: Ambeed
Warning

A150407-100MG A150407-250MG
103799-394EA 25 USD
103799-394 103804-162 77137-740 77137-742
(R)-5-(2,2-Dimethyl-4H-benzo[d][1,3]dioxin-6-yl)oxazolidin-2-one 97%
(R)-5-(2,2-Dimethyl-4H-benzo[d][1,3]dioxin-6-yl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₄
MW: 249.26 g/mol
Boiling Pt: 489.6±45.0 °C (760 mm Hg)
Flash Pt: 249.9 °C
Storage Temperature: Freezer
MDL Number: MFCD21362968
CAS Number: 452339-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR