(R)-(-)-4-Phenyl-2-oxazolidinone 97%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Phenyloxazolidin-2-one

A155508-100G A155508-10G A155508-1G A155508-500G A155508-25G A155508-5G
103842-968EA 6 USD
103842-968 76689-542 76689-544 76689-546 76689-548 77300-124 77318-694
(R)-(-)-4-Phenyl-2-oxazolidinone 97%
(R)-(-)-4-Phenyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 407 °C (760 mm Hg)
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00192393
CAS Number: 90319-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR