(R)-(+)-4-Isopropyl-2-oxazolidinone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-4-Isopropyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-2-oxazolidinone
A176198-1G
A176198-5G
A176198-10G
A176198-100G
103807-122EA
6
USD
103807-122
103845-442
77082-792
77082-794
77287-012
77748-150
(R)-(+)-4-Isopropyl-2-oxazolidinone 98%
(R)-(+)-4-Isopropyl-2-oxazolidinone
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Boiling Pt: 291.1 °C (760 mm Hg) Melting Pt: 70…73 °C Flash Pt: 129.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075172 CAS Number: 95530-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...