(R)-4-Boc-(3-hydroxymethyl)morpholine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Boc-(3-Hydroxymethyl)morpholine, tert-Butyl (3R)-3-(hydroxymethyl)morpholine-4-carboxylate

A281465-1G A281465-25G A281465-250MG A281465-5G
103813-550EA 20 USD
103813-550 103853-500 76837-198 76837-200 77276-982 77305-394
(R)-4-Boc-(3-hydroxymethyl)morpholine 97%
(R)-4-Boc-(3-hydroxymethyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
Boiling Pt: 320.7 °C (760 mm Hg)
Flash Pt: 147.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06799477
CAS Number: 215917-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR