(R)-4-Benzyl-3-(3-methylbutanoyl)oxazolidin-2-one 97%

Supplier: Ambeed
Warning

A143276-10G A143276-25G A143276-5G
103803-258EA 12 USD
103803-258 76719-404 76719-406 77316-510
(R)-4-Benzyl-3-(3-methylbutanoyl)oxazolidin-2-one 97%
(R)-4-Benzyl-3-(3-methylbutanoyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₃
MW: 261.32 g/mol
Boiling Pt: 412.9 °C (760 mm Hg)
Flash Pt: 203.5 °C
Storage Temperature: Ambient
MDL Number: MFCD10566718
CAS Number: 145589-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR