(R)-(+)-4-Benzyl-2-oxazolidinone 98%

Supplier: Ambeed
Warning

A175120-1000G A175120-100G A175120-10G A175120-500G A175120-25G A175120-5G
103845-308EA 7 USD
103845-308 76682-472 76682-474 76682-476 76682-478 77082-634 77082-636
(R)-(+)-4-Benzyl-2-oxazolidinone 98%
(R)-(+)-4-Benzyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
Boiling Pt: 398.8 °C (760 mm Hg)
Melting Pt: 87…90 °C
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00010846
CAS Number: 102029-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR