(R)-4-Amino-4-phenylbutanoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A134692-100MG A134692-250MG A134692-1G
103802-038EA 309 USD
103802-038 77149-782 77149-784 77298-718
(R)-4-Amino-4-phenylbutanoic acid hydrochloride 97%
(R)-4-Amino-4-phenylbutanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MW: 215.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12759018
CAS Number: 1010129-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR