(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-4-Boc-morpholine-3-carboxylic acid
A117398-100MG
A117398-10G
A117398-250MG
A117398-1G
A117398-5G
103798-880EA
37
USD
103798-880
103824-764
77136-332
77136-334
77136-336
77311-542
77311-544
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 95%
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₅ MW: 231.25 g/mol Boiling Pt: 369.5 °C (760 mm Hg) Melting Pt: 181 °C Flash Pt: 177.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114909 CAS Number: 869681-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...