(R)-4-(Iodomethyl)oxazolidin-2-one 95%

Supplier: Ambeed
Warning

A121867-100MG A121867-250MG
103800-148EA 180 USD
103800-148 76983-306 76983-308
(R)-4-(Iodomethyl)oxazolidin-2-one 95%
(R)-4-(Iodomethyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₄H₆INO₂
MW: 227 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18382539
CAS Number: 144542-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR