(R)-3-Phenylpyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

A280161-100MG A280161-25G A280161-5G A280161-250MG
103813-496EA 99 USD
103813-496 103853-440 77114-046 77305-352 77305-354 77593-730
(R)-3-Phenylpyrrolidine hydrochloride 97%
(R)-3-Phenylpyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClN
Boiling Pt: 280.5 °C (760 mm Hg)
Flash Pt: 123.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06796636
CAS Number: 851000-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR