(R)-3-Phenylpiperidine 97%

Supplier: Ambeed
Warning

A323022-100MG A323022-250MG
103825-320EA 377 USD
103825-320 76724-008 77601-448
(R)-3-Phenylpiperidine 97%
(R)-3-Phenylpiperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₅N
MW: 161.244 g/mol
Boiling Pt: 263.2 °C (760 mm Hg)
Melting Pt: 179…181 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839934
CAS Number: 430461-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR