(R)-3-Methylpiperidine hydrochloride 97%

Supplier: Ambeed
Warning

A316379-100MG A316379-250MG A316379-1G
103799-592EA 73 USD
103799-592 103825-098 103836-024 76875-802 76875-804 77306-144
(R)-3-Methylpiperidine hydrochloride 97%
(R)-3-Methylpiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClN
Storage Temperature: Ambient
MDL Number: MFCD22494946
CAS Number: 223792-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR