(R)-3-Methyl-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A177046-1G A177046-5G A177046-250MG
103807-206EA 458 USD
103807-206 103845-532 77082-910 77082-912 77163-754
(R)-3-Methyl-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butan-1-amine hydrochloride 95%
(R)-3-Methyl-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₄BNO₂·ClH
MW: 249.586 g/mol
Melting Pt: 198…201 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26403994
CAS Number: 1243174-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR