(R)-3-Methoxypyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A240836-100MG A240836-10G A240836-250MG A240836-1G A240836-5G
103811-366EA 23 USD
103811-366 103851-172 76786-518 76786-520 76786-522 77090-328 77090-330
(R)-3-Methoxypyrrolidine hydrochloride 95%
(R)-3-Methoxypyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD11849816
CAS Number: 474707-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR