(R)-(-)-3-Isopropyl-2,5-piperazinedione 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-3-Isopropylpiperazine-2,5-dione
A193943-250MG
A193943-25G
A193943-1G
A193943-5G
103816-600EA
25
USD
103816-600
103847-208
76769-948
76769-950
77085-110
77085-112
(R)-(-)-3-Isopropyl-2,5-piperazinedione 97%
(R)-(-)-3-Isopropyl-2,5-piperazinedione
Precaución: for Research Use Only
Formula:
C₇H₁₂N₂O₂ MW: 156.18 g/mol Boiling Pt: 404.3 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00210060 CAS Number: 143673-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...