(R)-3-Hydroxypyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Pyrrolidinol hydrochloride, (3R)-pyrrolidin-3-ol hydrochloride, (R)-3-Hydroxypyrrolidine HCl

A200586-100G A200586-10G A200586-500G A200586-25G A200586-5G
103847-848EA 9 USD
103847-848 76825-560 76825-562 76825-564 77302-694 77302-696
(R)-3-Hydroxypyrrolidine hydrochloride 97%
(R)-3-Hydroxypyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00191570
CAS Number: 104706-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR