(R)-3-Aminotetrahydrofuran hydrochloride 97%

Supplier: Ambeed
Warning

A393841-1G A393841-10G A393841-250MG
103826-930EA 9 USD
103826-930 103837-968 76841-378 77033-576 77033-578
(R)-3-Aminotetrahydrofuran hydrochloride 97%
(R)-3-Aminotetrahydrofuran hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.5800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08461720
CAS Number: 1072015-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR