(R)-3-Aminopyrrolidin-2-one 95%

Supplier: Ambeed
Warning

A113816-100MG A113816-250MG
103798-298EA 427 USD
103798-298 76971-046 76971-048
(R)-3-Aminopyrrolidin-2-one 95%
(R)-3-Aminopyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₄H₈N₂O
MW: 100.1191 g/mol
Boiling Pt: 305.6 °C (760 mm Hg)
Flash Pt: 138.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11501145
CAS Number: 121010-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR