(R)-(+)-3-Amino-1,2-propanediol 98%

Supplier: Ambeed
Danger

Synonyms: (R)-3-Aminopropane-1,2-diol, (R)-3-Amino-1,2-propanediol

A818353-1G A818353-25G A818353-10G A818353-5G
103841-118EA 30 USD
103841-118 76845-358 76845-360 76981-722 77308-376
(R)-(+)-3-Amino-1,2-propanediol 98%
(R)-(+)-3-Amino-1,2-propanediol

Precaución: For research use only

Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 266.7 °C (760 mm Hg)
Melting Pt: 54…56 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00798261
CAS Number: 66211-46-9
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR