D-β-Homoserine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Amino-4-hydroxybutanoic acid

A123435-250MG A123435-1G
103800-402EA 251 USD
103800-402 76995-956 77312-522
D-β-Homoserine 98%
D-beta-Homoserine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Boiling Pt: 374.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00270242
CAS Number: 16504-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR